Learn More
Agilent Technologies J&W CP-Sil 5 CB GC Column, 50 m, 0.32 mm, 0.40 μm, 7 inch cage. This is the most common GC column format compatible with Agilent 5890, 6890, 7820, 7890, 8860, and 8890 series GC systems and all non-Agilent GC systems with a similarly sized oven.
Supplier:  Agilent Technologies CP7749
Encompass
Capillary Tubing- Fused Silica, Film Thickness- 0.4 µm, Format- 7 inch, Inner Diameter (ID)- 0.32 mm, Length- 50 m, Phase- CP-Sil 5 CB, Polarity- Low Polarity, Temperature Range - -60 °C-325/350 °C, UNSPSC Code- 41115710, With Smart Key- No
Catalog No. NC3310949