Learn More
Medchemexpress LLC HY-101803 10mg Medchemexpress, CP671305 CAS:445295-04-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10180310MG
Encompass_Preferred
Medchemexpress, HY-101803 10mg CP671305 CAS:445295-04-5 CP671305 is a potent, orally active, selective inhibitor of phosphodiesterase-4-D, and possesses high activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7874