Learn More
Genetex Inc CPA1 Monoclonal Antibody [MSVA-601M] HistoMAX™ (GTX04433) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04433
Encompass_Preferred
The CPA1 antibody [MSVA-601M] HistoMAX™ (GTX04433) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human pancreas tissue using GTX04433 CPA1 antibody [MSVA-601M] HistoMAX. Strong CPA1 immunostaining in acinar cells of the pancreas.
Catalog No. NC3666941