Learn More
Cole-Parmer CPC (Colder) AseptiQuik W Quick-Disconnect Fitting, High-Flow Large Bore Genderless Hose Barb, Polycarbonate, 1-1/4" ID; 1/Ea
Supplier:  Cole-Parmer 5014364
Encompass_Preferred
High-flow, large-bore genderless sterilizable connectors simplify process integration while maximizing flexibility
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Robust construction provides reliable and repeatable fluid transfer
  • 1.5" nominal flow path size
  • 1-1/4" hose barb
  • The AseptiQuik genderless connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, simply unsnap and flip down the protective tab cover, align connectors with the pull tabs hanging down, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
    Catalog No. 50-006-10332