Learn More
Medchemexpress LLC Cpd-1224 | 2891620-68-9 | 99.8% | 855.40 | C43H47ClN8O7S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15338610MG
Encompass_Preferred
CPD-1224 is an orally active small-molecule inhibitor of anaplastic lymphoma kinase (ALK) that links an ALK-binding moiety to a cereblon ligand to induce target degradation. It degrades EML4-ALK and resistant mutant forms, shows tumor growth inhibition in preclinical studies, and is supplied for research applications.
  • Orally active anaplastic lymphoma kinase (ALK) inhibitor that promotes target degradation.
  • Effective against resistance mutations L1196M and G1202R.
  • High purity (>99.8%) suitable for analytical and biological assays.
  • Soluble in DMSO at approximately 14.29 mg/mL for in vitro studies.
  • Available in small-molecule quantities for preclinical research.
Catalog No. 50-002-86994