Learn More
Selleck Chemical LLC Cpd. 37 E4750-100MG
SDP
Supplier:  Selleck Chemical LLC E4750100MG
Encompass_Preferred
MYC-IN-3 (Cpd 37) is an alkynyl-substituted phenylpyrazole derivative and an inhibitor of MYC function by disrupting MYC/MAX interaction with an IC50 of 1 27 M in PC3 cells and preventing MYC/MAX binding to DNA It exhibits antiproliferative activity against multiple malignant cell lines and demonstrates enhanced therapeutic efficacy in a mouse allograft model of prostate cancer
Catalog No. 50-000-43634