Learn More
Medchemexpress LLC Cpd-39 | 3055472-38-0 | 98.1% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16866410MG
Encompass_Preferred
CPD-39 is a heterobifunctional PROTAC degrader that targets CCND1 and CDK4 and shows orally active anti-proliferative activity in preclinical studies. It is provided as a high-purity research reagent for laboratory investigations into targeted protein degradation and cell proliferation mechanisms.
  • Targets CCND1 and CDK4 for proteasomal degradation.
  • Demonstrates orally active anti-proliferative activity in preclinical models.
  • High purity, suitable for analytical and biological research.
  • Supplied in milligram-scale quantities for laboratory experiments.
  • Intended for research use only; not for human or diagnostic applications.
Catalog No. 50-002-95074