Learn More
Strem, An Ascensus Company CAS# 1447963-73-6. 100mg. Methanesulfonato(2-dicyclohexylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), 98% [CPhos Palladacycle Gen. 3].
SDP
Supplier:  Strem, An Ascensus Company 460487100MG
Encompass
CAS# 1447963-73-6. 100mg. Methanesulfonato(2-dicyclohexylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), 98% [CPhos Palladacycle Gen. 3]. . Molecular Weight: 806.35. Molecular Formula: C41H54N3O3PPdS. Color/form: white to off-white pwdr. Strem# 46-0487. http://www.strem.com/catalog/v/46-0487/
Catalog No. 50-201-6705