Learn More
TARGETMOL CHEMICALS INC CPI1189 10mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T356110MG
Encompass_Preferred
REN-1189 is a TNF-a release inhibitor potentially for the treatment of sciatica and postherpetic neuralgia.
Catalog No. NC1624935