Learn More
TARGETMOL CHEMICALS INC CPI-1205 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T85105MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CPI-1205 is a highly potent and selective inhibitor of EZH2(IC50 0.002 uM EC50 0.032 uM) . purity: 99%
Catalog No. 50-257-9063