Learn More
Medchemexpress LLC CPI-637 | 1884712-47-3 | 99.9% | 386.45 | C22H22N6O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100482100MG
Encompass_Preferred
CPI-637 is a selective, cell-active CBP/EP300 bromodomain inhibitor that exhibits low-nanomolar biochemical potency and confirmed cellular activity. It inhibits CBP and EP300 with IC50 values in the tens of nanomolar range, shows substantially weaker activity against BRD4 BD-1, and has reported cellular EC50 and antiproliferative effects in multiple cancer cell lines. It is supplied as a high-purity research compound for epigenetics and oncology studies.
  • Selective CBP/EP300 bromodomain inhibition.
  • Low-nanomolar biochemical potency.
  • Demonstrated cellular activity and EC50 measurements.
  • Antiproliferative effects reported in multiple cancer cell lines.
  • High chemical purity suitable for research.
  • Supplied as a 100 mg research pack.
Catalog No. 50-002-13964