Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CELLULAR TECHNOLOGY LTD CPI Positive Control Solution, 0.5 ml, 80x concentration, 20x concentration, frozen protein pool solution
SDP

Supplier:  CELLULAR TECHNOLOGY LTD CTLCPI005

Encompass

The CPI Positive Control Solution is a ready-to-use, frozen formulation that serves as a universal positive control for assessing the functionality of antigen-specific CD4+ T cells and Antigen Presenting Cells (APCs) in human peripheral blood mononuclear cells (PBMC). This product is essential for immunological assays such as IFN-γ ELISPOT and ELISA, containing an inactivated protein pool of Cytomegalovirus, Parainfluenza, and Influenza viruses to stimulate antigen-specific responses.

  • Contains an inactivated protein pool for optimal CD4+ T cell stimulation.
  • Compatible with multiple cytokine assays including ELISPOT and ELISA.
  • Sterile filtered and rigorously tested for performance.
  • Recommended dilutions of 1:80 and 1:20; 50 μg/ml.
  • Storage at 4°C with a shelf life of six months or -20°C for one year.
  • Tested with ImmunoSpot Platform for highest quality assurance.

Catalog No. NC2330015


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.