Learn More
Trajan Scientific CPLRY 25QC3/BP20-0.5
SDP
Supplier:  Trajan Scientific 054436
Encompass_Preferred
SGE BP20 (Wax) GC Capillary Columns, Diameter Inner: 0.32 mm, Film Thickness: 0.5 um, Length: 25 m, For Use With: Free Acids, Solvents and Essential Oils, Aromatics Analysis, Alcohol Analysis, Fatty Acid Methyl Esters, Packing Material: SGE BP20 (Wax), Stationary Phase: Polyethylene Glycol
Catalog No. 50-253-0238