Learn More
Trajan Scientific CPLRY SOLGEL-WAX-30X.32X.50
SDP
Supplier:  Trajan Scientific 054797
Encompass_Preferred
Trajan SolGel-WAX GC Capillary Columns, Diameter Inner: 0.32 mm, Film Thickness: 0.5 um, Length: 30 m, Particle Size: 0.5 um, For Use With: Highly Active Compounds Analysis, Packing Material: SGE SolGel-Wax, Stationary Phase: Polyethylene Glycol, Polarity: Polar
Catalog No. 50-253-0231