Learn More
AdipoGen CPM
SDP
Supplier:  AdipoGen CDXD0042M250
Encompass_Preferred
Chemical. CAS 76877-33-3. Formula C24H22N2O4. MW 402.44. Synthetic. Widely used blue fluorescent thiol-reactive dye Ex/Em 384/470nm. This maleimide derivative of coumarin is essentially non-fluorescent until it reacts with thiols, making it possible to quantify thiols without a separation step. Good energy acceptor from tryptophan and a good energy donor to fluorescein. Used to monitor release of thiols and to distinguish proliferating cancer cells by nucleolar protein staining.
Catalog No. 50-256-1288