Learn More
Medchemexpress LLC Cpn-219 | 2240202-02-0 | 99.08% | 849.08 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP303010MG
Encompass_Preferred
CPN-219 is a next-generation hexapeptidic NMUR2 agonist. It achieves dose-dependent and selective activation of NMUR2 with an EC50 value of 2.2 nM. For research purposes, CPN-219 has shown an ability to suppress body weight gain in mice through intranasal administration in a dose-dependent manner.
  • Acts as a next-generation hexapeptidic NMUR2 agonist
  • Achieves dose-dependent and selective activation of NMUR2
  • EC50 value of 2.2 nM
  • Suppresses body weight gain in mice via intranasal administration
  • Ineffective via intraperitoneal injection in mice at 250 μg
Catalog No. 50-004-00093