Learn More
TARGETMOL CHEMICALS INC CPPHA 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1088125MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CPPHA a positive allosteric modulator of glutamate receptors mGluR5 and mGluR1 is commonly used in development for central nervous system diseases. purity: 99%
Catalog No. 50-258-1630