Learn More
Cayman Chemical CPS1NTerm RabbIt Monclon 100uL
SDP
Supplier:  Cayman Chemical 32320100 UL
Encompass_Preferred
A pyrimidine analog and nitrosourea alkylating agent inhibits the growth of U-251MG and EA285 glioma cells at 10 g/ml increases the median survival time in a Walker 256 carcinoma rat model of metastatic brain tumors at 30 mg/kg
Catalog No. 50-294-7817