Learn More
Medchemexpress LLC (±)-CPSI-1306 | 1309793-47-2 | 99.64% | 310.30 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY110095
Encompass_Preferred
(±)-CPSI-1306 is an orally available antagonist of macrophage migration inhibitory factor (MIF). It is for research use only.
  • Orally available MIF antagonist.
  • Shows a significant drop in blood glucose levels and a reduction in serum levels of inflammatory cytokines in mice.
  • Effective in treating non-insulin-dependent diabetes mellitus in mice.
  • Induces keratinocyte apoptosis as early as 30 minutes after single UVB exposure.
  • Reduces acute UVB-induced keratinocyte DNA damage and UVB-induced acute inflammation.
Catalog No. 50-003-47574