Learn More
Medchemexpress LLC Cpth2-alkyne | 1613116-16-7 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1203171MG
Encompass_Preferred
Alkyne-tagged analog of a NAT10 inhibitor used as a cell-active chemical probe to study nuclear architecture and laminopathy. The compound localizes to nucleoli, nuclear periphery, and cytoplasm, and is used for cell-based localization and target engagement studies.
  • Alkyne tag for bioorthogonal chemistry applications.
  • Cell active and accumulates in nucleoli.
  • Inhibits N-acetyltransferase 10 (NAT10).
  • High reported purity suitable for biochemical assays.
  • Available in a 1 mg research quantity.
Catalog No. 50-002-31696