Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CPTH2-alkyne | 1613116-16-7 | 99.0% | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1203175MG

Encompass_Preferred

CPTH2-alkyne is an alkyne-tagged analog of CPTH2 that inhibits N-acetyltransferase 10 (NAT10). It accumulates in nucleoli, localizes to the nuclear periphery and cytoplasm, and is supplied as a high-purity research compound for studies of nuclear architecture and laminopathy.

  • Alkyne-tagged analog suitable for bioorthogonal click chemistry.
  • Inhibits N-acetyltransferase 10 (NAT10) for epigenetic studies.
  • Accumulates in nucleoli and localizes to nuclear periphery and cytoplasm.
  • High purity (99.0%) for research applications.
  • Stable as a powder at -20°C for up to 3 years.

Catalog No. 50-002-34412


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.