Learn More
ABclonal Technology CPVL Rabbit pAb
SDP
Supplier:  ABclonal Technology A123691000UL
Encompass_Preferred
The protein encoded by this gene is a carboxypeptidase and bears strong sequence similarity to serine carboxypeptidases. Carboxypeptidases are a large class of proteases that act to cleave a single amino acid from the carboxy termini of proteins or peptides. The exact function of this protein, however, has not been determined.
Catalog No. 50-000-99348