Learn More
Medchemexpress LLC CPYPP is a DOCK2-Rac1 interaction inhibitor. | 310460-39-0 | 98.5% | 324.76 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY110100100MG
Encompass_Preferred
CPYPP is a DOCK2-Rac1 interaction inhibitor that binds to the DOCK2 DHR-2 domain and inhibits the guanine nucleotide exchange factor (GEF) activity of DOCK2DHR-2 for Rac1. It also inhibits DOCK180 and DOCK5, with less significant inhibition of DOCK9. This compound has been shown to block chemokine receptor- and antigen receptor-mediated Rac activation in lymphocytes, leading to reduced chemotactic response and T cell activation. In HEK293T cells, CPYPP markedly suppresses DOCK2-induced Rac activation.
  • Binds to the DOCK2 DHR-2 domain
  • Inhibits GEF activity of DOCK2DHR-2 for Rac1
  • Inhibits DOCK180 and DOCK5
  • Blocks chemokine and antigen receptor-mediated Rac activation
  • Reduces chemotactic response and T cell activation
  • Suppresses DOCK2-induced Rac activation
Catalog No. 50-003-44128