Learn More
Medchemexpress LLC HY-136453 5mg Medchemexpress, CR-1-31-B CAS:1352914-52-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1364535MG
Encompass_Preferred
Medchemexpress, HY-136453 5mg CR-1-31-B CAS:1352914-52-3 CR-1-31-B is a potent eIF4A RNA helicase inhibitor. CR-1-31-B blocks MUC1-C translation in response to growth factor stimulation in breast cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0442