Learn More
Medchemexpress LLC Crac intermediate 1 | 1249343-86-9 | 98.5% | 10 MG

Supplier: Medchemexpress LLC HY2058710MG
CRAC intermediate 1 is a synthetic chemical intermediate used in the laboratory synthesis of CRAC channel inhibitors. It is supplied for research use only and is described in patent literature as a key intermediate in the synthesis pathway.
- CAS number: 1249343-86-9.
- Purity: 98.5%.
- Molecular formula: C10H7F2N3O.
- Molecular weight: 223.18 g/mol.
- Appearance: white to off-white solid.
- Solubility: ≥50 mg/mL in DMSO.
- Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (2 years).
- Intended use: for research use only, not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.