Learn More
ABclonal Technology CRACR2A Rabbit pAb
SDP
Supplier:  ABclonal Technology A138391000UL
Encompass_Preferred
Enables GTPase activity and calcium ion binding activity. Involved in several processes, including activation of store-operated calcium channel activity, positive regulation of JNK cascade, and store-operated calcium entry. Located in several cellular components, including Golgi apparatus, Weibel-Palade body, and immunological synapse.
Catalog No. 50-000-98511