Learn More
Medchemexpress LLC CRAMP (mouse) | 376364-36-2 | >99.0% | 3878.61 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP485510MG
Encompass_Preferred
CRAMP (mouse) is an antimicrobial peptide, also known as Cathelicidin-related antimicrobial peptide, and is a functional homolog of LL-37. It exhibits potent antibacterial activity, particularly against Gram-negative bacteria. This peptide is also involved in various biological processes including wound healing, immune regulation, and the maintenance of intestinal homeostasis.
  • Exhibits potent antibacterial activity against Gram-negative bacteria.
  • Can activate macrophages to secrete TNF-α when complexed with CpG.
  • Involved in wound healing.
  • Plays a role in immune regulation.
  • Contributes to the maintenance of intestinal homeostasis.
  • Forms complexes with bacterial genomic DNA in vitro.
Catalog No. 50-003-98592