Learn More
Cayman Chemical CrasIcaulIn A 50mg
SDP
Supplier:  Cayman Chemical 3467450 MG
Encompass_Preferred
A diterpene alkaloid with analgesic activity; inhibits acetic acid-induced writhing in mice (ED50 = 0.048 mg/kg); induces lethality in mice at >0.92 mg/kg
Catalog No. 50-294-6265