Learn More
Electron Microscopy Sciences AMB Passive Air Platform Shipping Crate
SDP
Supplier:  Electron Microscopy Sciences 6670
Encompass_Preferred
These high performance anti vibration platform for balances are weighted to 25kg and are for use with light instruments such as balances. They support loads to 120kg and are available in three sizes, 45x30cm, 40x50cm and 60x60cm. Their small footprint also makes them ideal for use when space is at a minimum. Working height is 6cm.
Catalog No. 50-344-72