Learn More
Enterprise Technology Solutions Crayola Paint, Acrylic, 16 oz, Ivory Black
SDP
Supplier:  Enterprise Technology Solutions CYO204016244
Encompass_Preferred
Light up your art with Portfolio Series Acrylic Paint. Ideal for crafts at school or home, this paint for crafting features a creamy consistency and opaque finish that won't chip or flake. Acrylic paint can be used on almost any surface making it great for arts and crafts, school projects, classroom activities, DIY crafts and more. It is great for teaching young artists how to mix, blend and create color. Rich, vivid colors are so pigment-enriched they can be diluted 8 to 1 for watercolor techniques without losing color intensity. Also use for applications as varied as airbrushing, printmaking and marbling without chemical additive. Nontoxic, child-safe paint is ideal for children ages 3 and up.
Catalog No. 50-008-26552