Learn More
Medchemexpress LLC CRBN ligand-11 | 2962967-27-5 | 99.9% | 301.11 | 10 G
SDP
Supplier:  Medchemexpress LLC HY16998910G
Encompass_Preferred
CRBN ligand-11 is an E3 ubiquitin ligase ligand for CFT8634 and can be used for the synthesis of PROTACs.
  • E3 ubiquitin ligase ligand.
  • Used for the synthesis of PROTACs.
  • Targets cereblon.
  • Solid, white to off-white appearance.
  • Soluble in DMSO (100 mg/mL).
Catalog No. 50-003-91330