Learn More
Medchemexpress LLC Creatine-13C | 687996-99-2 | 98.5% | 132.13 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW010388S5MG
Encompass_Preferred
Creatine-13C is the 13C labeled Creatine, an endogenous amino acid derivative important for cellular energy, particularly in muscle and brain. This compound is for research use only and not intended for human use.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, like 13C, are incorporated into drug molecules to aid in quantitation during the drug development process.
Catalog No. 50-004-53894