Learn More
Selleck Chemical LLC Creatine
SDP
Supplier:  Selleck Chemical LLC S55881G
Encompass_Preferred
Creatine (Methylguanidoacetic acid) is a nitrogenous organic acid that occurs naturally in vertebrates It facilitates the recycling of adenosine triphosphate (ATP) primarily in muscle and brain tissue Creatine can inhibits the JAK-STAT1 signal transmission by inhibiting the interaction of IFN- receptors with JAK2 in an ATP-independent manner thereby inhibiting downstream pro-inflammatory gene expression
Catalog No. 50-000-03154