Learn More
Medchemexpress LLC Creatine-d3 | 143827-19-4 | 99.5% | 134.15 | 5 MG
Supplier: Medchemexpress LLC HYW010388S15MG
Creatine-d3 is a deuterium-labeled Creatine, an endogenous amino acid derivative crucial for cellular energy, particularly in muscle and brain. Deuteration, the process of incorporating stable heavy isotopes into drug molecules, is largely used as a tracer for quantitation during drug development. This product can be used as a tracer and as an internal standard for quantitative analysis.
- Used as a tracer for quantitation during drug development
- Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
- Plays an important role in cellular energy, especially in muscle and brain
- Stable isotope-labeled compound