Learn More
Medchemexpress LLC Creatine-d3 hydrate | 284664-86-4 | 99.0% | 152.17 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW010388AS1MG
Encompass_Preferred
Creatine-d3 (hydrate) is a deuterium-labeled Creatine hydrate. Creatine hydrate is essential in the energy metabolism of muscle and brain cells under both physiological and pathological conditions.
  • Used as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-67068