Learn More
Bioworld Creatine Phosphate, 25 g
Supplier:  Bioworld 403001122
Encompass_Preferred
A preparation of creatine Specifications:Also known as Phosphocreatine Disodium HydrateHigh Purity GradeMoisture: <25.0%ATP (enzymatic): >0.001%Inorganic Phosphate: <0.01%Appearance: Solid
Catalog No. 50-255-680