Learn More
Cayman Chemical CreatInIn ELISA KIt 96Wel
SDP
Supplier:  Cayman Chemical 50233096 WELL
Encompass_Preferred
Cayman’s Creatinine ELISA Kit is a competitive assay that can be used for the quantification of creatinine in plasma, serum, and urine. The assay has a range of 0.273-35 UG/ml, with a midpoint (50% B/B0) of 2.5-5.5 UG/ml, and a sensitivity (80% B/B0) of approximately 1 UG/ml.
Catalog No. 50-289-3954