Learn More
Cambridge Isotope Laboratories Creatinine (N-Methyl-D3, 98%), 143827-20-7, MFCD00190410, 0.1G
Supplier:  Cambridge Isotope Laboratories DLM36530.1
Encompass
Chemical Formula: C4H4D3N3O, Molecular Weight: 116.14, Synonym: 4-Amidinosarcosine-d3 (methyl-d3), MP: 255 °C (dec.) (lit.)
Catalog No. NC1407593