Learn More
Creative Biolabs Creative Biolabs Recombinant Antibody, Mouse, Anti-APP, clone 3D6, Human

Supplier: Creative Biolabs PABL01150UG
This recombinant mouse antibody (3D6) specifically binds to APP (amyloid beta precursor protein). It is a neo-epitope specific antibody that recognizes amyloid-β1-5 with a strong preference for an exposed Asp residue at the N-terminus of the peptide.
- Immunogen: Human amyloid beta precursor protein
- Host species: Mouse
- Type: Mouse IgG1
- Specificity: Human APP
- Species reactivity: Human
- Clone: 3D6
- Applications: Western Blot, Enzyme-linked Immunosorbent Assay, Functional Assay
- Purity: >95% as determined by analysis by SDS-PAGE
- Buffer: PBS
- Preservative: No preservatives
- Storage: Centrifuge briefly prior to opening vial. Store at +4°C short term (1 to 2 weeks). Aliquot and store at −20°C long term. Avoid repeated freeze/thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.