Learn More
Medchemexpress LLC Crebinostat 10mg | 1092061-61-4 | 353.41 g·mol⁻¹ | C20H23N3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11681810MG
Encompass_Preferred
Crebinostat is a small-molecule histone deacetylase (HDAC) inhibitor for research use. It targets multiple HDAC isoforms, induces acetylation of histone H3 and H4, and has been used in preclinical studies to modulate neuroplasticity and memory.
  • Potent inhibitor of HDAC1, HDAC2, HDAC3, and HDAC6.
  • Induces histone H3 and H4 acetylation in cellular assays.
  • Demonstrated effects on chromatin-mediated neuroplasticity and memory in animal models.
  • Available as a high-purity research powder (10 mg size available).
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-27685