Learn More
Medchemexpress LLC Crebinostat 25mg | 1092061-61-4 | 353.41 g/mol | C20H23N3O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11681825MG
Encompass_Preferred
Crebinostat is a small-molecule histone deacetylase (HDAC) inhibitor provided as a solid powder for research use. It inhibits class I HDACs and HDAC6, increases histone acetylation, and is used in preclinical studies of epigenetic regulation, neurobiology, and oncology.
  • Potent inhibition of HDAC1, HDAC2, HDAC3, and HDAC6
  • High purity suitable for biochemical and cell-based assays
  • Stable as a powder under recommended storage conditions
  • Available in multiple small-scale package sizes for research workflows
Catalog No. 50-002-28984