Learn More
Medchemexpress LLC Crebinostat 50mg | 1092061-61-4 | 50 MG

Supplier: Medchemexpress LLC HY11681850MG
Crebinostat is a small-molecule histone deacetylase (HDAC) inhibitor used in biochemical and cellular research to probe epigenetic regulation. It is supplied as a purified solid for laboratory research use only.
- Potent HDAC inhibitor (IC50: HDAC1 0.7 nM; HDAC2 1.0 nM; HDAC3 2.0 nM; HDAC6 9.3 nM).
- High purity: 99.79%.
- Molecular formula C20H23N3O3 and molecular weight 353.41 g/mol.
- Supplied as a solid suitable for in vitro and preclinical studies; follow SDS guidance for handling.
- Available in multiple milligram quantities, including 50 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.