Learn More
Medchemexpress LLC Crebinostat 5mg | 1092061-61-4 | 353.41 | C20H23N3O3 | 5 MG

Supplier: Medchemexpress LLC HY1168185MG
Crebinostat is a small-molecule histone deacetylase (HDAC) inhibitor supplied for research use. It is provided in milligram quantities for in vitro and preclinical studies involving epigenetic modulation and cell-cycle/DNA damage pathways.
- Potent HDAC inhibitor for epigenetic and cell cycle research.
- Provided in small milligram pack sizes for convenient dosing.
- Suitable for in vitro and preclinical applications.
- Molecular formula C20H23N3O3 and molecular weight 353.41.
- CAS 1092061-61-4 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.