Learn More
Sino Biological Peptide Substrates: CREBtide
Supplier:  Sino Biological C5058
Encompass_Preferred
The CREBtide peptide sequence (KRREILSRRPSYR) is based on the human CREB1 isoform A (amino acid 109-121).
Catalog No. 50-256-9521