Learn More
Selleck Chemical LLC CREBtide P1155-5MG
SDP
Supplier:  Selleck Chemical LLC P11555MG
Encompass_Preferred
CREBtide a synthetic 13 amino acid peptide has been reported as a PKA substrate
Catalog No. 50-000-43623