Learn More
Selleck Chemical LLC Crenigacestat
SDP
Supplier:  Selleck Chemical LLC S716925MG
Encompass_Preferred
Crenigacestat (LY3039478) is an oral Notch and gamma-secretase inhibitor with IC50 of 0 41 nM for Notch
Catalog No. 50-000-04176