Learn More
TARGETMOL CHEMICALS INC CRENOLANIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T267710MG
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Crenolanib (ARO 002) is an orally bioavailable type III tyrosine kinases inhibitor of PDGFR(alpha)/(beta) and FLT3 (IC50s 11 3.2 and 4 nM). purity: 99%
Catalog No. 50-257-7063