Learn More
Medchemexpress LLC Crenolanib (benzenesulfonate) | 670220-93-6 | MFCD31693693 | 99.5% | 601.72 g/mol | C32H35N5O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13223A5MG
Encompass_Preferred
Crenolanib (benzenesulfonate) is the besylate salt form of crenolanib, a potent and selective inhibitor of class III receptor tyrosine kinases (FLT3 and PDGFRα/β). Supplied as a solid research reagent in small mass quantities, it is used for in vitro studies of kinase signaling and assay development. The material has a high reported purity and defined storage recommendations.
  • High purity (99.45%) suitable for research applications.
  • Solid powder form for accurate dosing and storage.
  • Potent, selective inhibition of FLT3 and PDGFR signaling.
  • Available in small research sizes for assay development.
  • Recommended storage: 4°C sealed; in solution store at -80°C for long-term.
Catalog No. 50-002-62594