Learn More
MARSHALL SCIENTIFIC Crest P1200HTPC-45 Digital Ultrasonic Cleaner w/ Power Control
SDP
Supplier:  MARSHALL SCIENTIFIC 1200PD0451
Encompass_Preferred
The Crest P1200HTPC-45 Digital Ultrasonic Cleaner w/ Power Control has a 2.5 gallon tank size and utilizes a digital heater to set temperature and total run time.
Catalog No. NC2591244