Learn More
Aat Bioquest Cresyl violet *CAS 41830-80-2*
SDP
Supplier:  Aat Bioquest 90
Encompass_Preferred
It is used for staining Nissl substances in nerve cells and for bulk staining of nerve tissue. It is also used for preparing Magic Red-based protease substrates that have red fluorescence upon enzyme hydrolysis.
Catalog No. 50-195-2705